Orion™ Single Junction Combination pH Electrodes
Thermo Scientific™

Orion™ Single Junction Combination pH Electrodes

Confidently measure your samples with the Orion Silver Chloride pH electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesJunctionTypeMaterial
9146BNBNCWickCombination pH ElectrodeEpoxy Body
9142BNBNCCeramicCombination pH ElectrodeGlass Body
9147BNRequires BNC-to-USS AdapterHDPE pinpH/ATC Combination ElectrodeEpoxy Body
Catalog number 9146BN
Price (USD)
101.70
Online exclusive
113.00
Save 11.30
Each
Connector Types:
BNC
Junction:
Wick
Type:
Combination pH Electrode
Material:
Epoxy Body
Price (USD)
101.70
Online exclusive
113.00
Save 11.30
Each
Thermo Scientific™ Orion™ Silver Chloride pH Electrodes are available in different body materials with either refillable or low maintenance gel options too. The 9142BN is a refillable electrode with chemically resistant glass body best suited for general purpose samples, including solvents, while the 9146BN and 9147BN contain a low-maintenance gel that is best suited for field or plant use, and also educational institutes, where minimal maintenance is preferred. The 9147BN also comes with a built-in ATC.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
DescriptionLow maintenance gel combination with epoxy body and fluted guard
For Use With (Equipment)Russell meters
Storage RequirementsStore in Orion pH electrode storage solution 910001
Temperature (English) Operating32°F to 176°F
Tip TypeRemoveable Bulb Guard
Connector TypesBNC
JunctionWick
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 80°C
TypeCombination pH Electrode
pH Range0 to 14
Unit SizeEach