Orion™ Single Junction Combination pH Electrodes
Thermo Scientific™

Orion™ Single Junction Combination pH Electrodes

Confidently measure your samples with the Orion Silver Chloride pH electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesJunctionTypeMaterial
9147BNRequires BNC-to-USS AdapterHDPE pinpH/ATC Combination ElectrodeEpoxy Body
9142BNBNCCeramicCombination pH ElectrodeGlass Body
9146BNBNCWickCombination pH ElectrodeEpoxy Body
Catalog number 9147BN
Price (USD)
177.30
Online exclusive
197.00
Save 19.70
Each
Connector Types:
Requires BNC-to-USS Adapter
Junction:
HDPE pin
Type:
pH/ATC Combination Electrode
Material:
Epoxy Body
Price (USD)
177.30
Online exclusive
197.00
Save 19.70
Each
Thermo Scientific™ Orion™ Silver Chloride pH Electrodes are available in different body materials with either refillable or low maintenance gel options too. The 9142BN is a refillable electrode with chemically resistant glass body best suited for general purpose samples, including solvents, while the 9146BN and 9147BN contain a low-maintenance gel that is best suited for field or plant use, and also educational institutes, where minimal maintenance is preferred. The 9147BN also comes with a built-in ATC.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
DescriptionLow maintenance gel combination w/epoxy body, built-in ATC
For Use With (Equipment)Russell meters
Storage RequirementsStore in Orion pH electrode storage solution 910001
Temperature (English) Operating32°F to 122°F
Connector TypesRequires BNC-to-USS Adapter
JunctionHDPE pin
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 50°C
TypepH/ATC Combination Electrode
pH Range0 to 14
Unit SizeEach