Orion™ Single Junction Refillable Epoxy Bodied Combination pH Electrode
Orion™ Single Junction Refillable Epoxy Bodied Combination pH Electrode
Orion™ Single Junction Refillable Epoxy Bodied Combination pH Electrode
Thermo Scientific™

Orion™ Single Junction Refillable Epoxy Bodied Combination pH Electrode

Confidently maneuver and measure in the field or on the plant floor with Orion™ refillable epoxy body electrodes.
Have Questions?
Catalog NumberConnector Types
9156BNWPBNC (Waterproof)
Catalog number 9156BNWP
Price (USD)
218.70
Online exclusive
243.00
Save 24.30
Each
Connector Types:
BNC (Waterproof)
Price (USD)
218.70
Online exclusive
243.00
Save 24.30
Each
Thermo Scientific™ Orion™ Silver Chloride Refillable Electrodes provide reliable and accurate measurements of routine samples. They are available is epoxy bodies for ruggedness and best suited for general purpose samples and drinking water.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
DescriptionGeneral purpose combination pH electrode with epoxy body and waterproof BNC connection
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
Filling Solution Type4M KCl with Ag/AgCl
For Use With (Equipment)Standard pH Meter
Internal ReferenceAg/AgCl
Length (English)4.7 in.
Length (Metric)120 mm
Precision0.02
Temperature (English) Operating32°F to 194°F
Connector TypesBNC (Waterproof)
JunctionGlass Fiber
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 90°C
TypeCombination pH Electrode, BNC waterproof connector
pH Range0 to 14
Unit SizeEach