96-Well Microtiter™ Microplates
Actual product may vary
96-Well Microtiter™ Microplates
96-Well Microtiter™ Microplates
Thermo Scientific™
96-Well Microtiter™ Microplates
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialWell ShapeSterilityVolume (Metric) Well
9205PolystyreneCurved Edges, Flat Bottom (C-well)Non-sterile330 μL
2305-11PolystyreneRound Bottom (U-well)Sterile280 μL
2205PolystyreneRound Bottom (U-well)Non-sterile400 μL
2605PolystyreneConical Bottom (V-well)Non-sterile280 μL
Catalog number 9205
Price (USD)
170.00
Case of 50
Material:
Polystyrene
Well Shape:
Curved Edges, Flat Bottom (C-well)
Sterility:
Non-sterile
Volume (Metric) Well:
330 μL
Price (USD)
170.00
Case of 50
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
  • For high-throughput screening and research
  • Clear plates for colorimetric applications
  • Standard footprint, 128 x 86 mm
  • Available as both polystyrene square wells and vinyl square wells
  • Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
  • Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
FormatStandard
LidWithout Lid
No. per Case50/Cs.
No. per Pack10 Pk.
Packaging50 Cases, 10 Pk.
Product TypeMicroplate
Quantity50/Pk.
Volume (Metric) Well330 μL
Array8 x 12
BarcodeNo Barcode
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineMicrotiter
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeCurved Edges, Flat Bottom (C-well)
Unit SizeCase of 50
Frequently asked questions (FAQs)
What is the well shape and well capacity for 96-Well Microtiter Microplates (Cat. No. 9205)? Do they have a lid?
96-Well Microtiter Microplates (Cat. No. 9205) do not have a lid and have the following well shape: curved edges, flat bottom (C-well). Each well can hold 330 µL volume.