Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
Thermo Scientific™
Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
Increase pipetting effectiveness with these automation tips, achieving accurate and precise performance due to superior tip straightness and a low coefficient of variation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)AutoclavableFilteredFit for PurposeUnit Size
929-262PK
also known as 929-262
235 μLAutoclavableNon-filteredCertified QualityPack of 960
925-02110 μLNot AutoclavableFilteredSterile and PCR ReadyCase of 4800
925-26120 μLNot AutoclavableNon-filteredSterileCase of 4800
925-261PK
also known as 925-261
20 μLNot AutoclavableNon-filteredSterilePack of 960
925-26220 μLAutoclavableNon-filteredCertified QualityCase of 4800
927-02140 μLNot AutoclavableFilteredSterile and PCR ReadyCase of 4800
927-021PK
also known as 927-021
40 μLNot AutoclavableFilteredSterile and PCR ReadyPack of 960
927-26150 μLNot AutoclavableNon-filteredSterileCase of 4800
927-261PK
also known as 927-261
50 μLNot AutoclavableNon-filteredSterilePack of 960
927-26250 μLAutoclavableNon-filteredCertified QualityCase of 4800
927-262PK
also known as 927-262
50 μLAutoclavableNon-filteredCertified QualityPack of 960
929-021180 μLNot AutoclavableFilteredSterile and PCR ReadyCase of 4800
929-021PK
also known as 929-021
180 μLNot AutoclavableFilteredSterile and PCR ReadyPack of 960
929-261235 μLNot AutoclavableNon-filteredSterileCase of 4800
929-261PK
also known as 929-261
235 μLNot AutoclavableNon-filteredSterilePack of 960
929-262235 μLAutoclavableNon-filteredCertified QualityCase of 4800
931-02120 μLNot AutoclavableFilteredSterile and PCR ReadyCase of 19,200
927-261-0550 μLNot AutoclavableNon-filteredSterileCase of 4800
Catalog number 929-262PK
also known as 929-262
Price (USD)
150.75
Online Exclusive
167.50
Save 16.75 (10%)
Pack of 960
Volume (Metric):
235 μL
Autoclavable:
Autoclavable
Filtered:
Non-filtered
Fit for Purpose:
Certified Quality
Unit Size:
Pack of 960
Price (USD)
150.75
Online Exclusive
167.50
Save 16.75 (10%)
Pack of 960
Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty: A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
Packaging96 tips/rack, 10 racks/pack, 5 packs/case
Quantity4800 tips
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Packard Liquid Handling System, PerkinElmer
FormatRack
Product LinePerkinElmer/Packard
SterilityNon-sterile
Tip StyleAutomation
Volume (Metric)235 μL
Unit SizePack of 960