Which version of Chromosome Analysis Suite (ChAS) Software is required to analyze CytoScan XON arrays?
Chromosome Analysis Suite (ChAS) Software version 3.3 is required to analyze CytoScan XON arrays.