Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFilteredFit for PurposeUnit Size
935-261SterileNon-filteredSterileCase of 19200
935-261PK
also known as 935-261
SterileNon-filteredSterilePack of 3840
935-262Non-sterileNon-filteredCertified QualityCase of 19200
935-262PK
also known as 935-262
Non-sterileNon-filteredCertified QualityPack of 3840
Catalog number 935-261
Price (USD)
3,227.50
Online exclusive
6,455.00
Save 3,227.50
Case of 19200
Sterility:
Sterile
Filtered:
Non-filtered
Fit for Purpose:
Sterile
Unit Size:
Case of 19200
Price (USD)
3,227.50
Online exclusive
6,455.00
Save 3,227.50
Case of 19200
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
Product LineBeckman
SterilitySterile
Tip StyleExtended Length
Volume (Metric)30 μL
Unit SizeCase of 19200