Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Thermo Scientific™

Finntip™ Non-Filtered 384 Format Pipette Tips 20 μL

Ensure accurate, precise pipetting. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFit for PurposeFormat
9400613SterileSterile & PCR ReadyRack
9400620Non-sterileCertified QualityBulk
9400610Non-sterileCertified QualityRack
Catalog number 9400613
Price (USD)
673.03
Online exclusive
798.00
Save 124.97
Case of 3840
Sterility:
Sterile
Fit for Purpose:
Sterile & PCR Ready
Format:
Rack
Price (USD)
673.03
Online exclusive
798.00
Save 124.97
Case of 3840

Accurate, precise pipetting with 384-format volume 20 μL. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Key Features:

  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack

Proven quality

  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Immediate traceability

  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options

Note:

Availability may vary by country.

Specifications
DescriptionFinntip 20 Sterile, Non-Filtered, Sterile, Rack
Length (English)1.3 in.
Length (Metric)3.3cm
ModelFinntip 20 Sterile
Shelf LifeRecommended shelf life for Finntip pipette tips is five years
AutoclavableFinntip pipette tips and racks can be autoclaved for 121 degrees Celsius for 20 minutes
ColorPurple
Color CodePurple
FilteredNon-filtered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Thermo Scientific Finnpipette 20
FormatRack
Product LineFinnTip
SterilitySterile
Volume (Metric)20 μL
Unit SizeCase of 3840