(±)-1,2-Propanediol, ≥99.5%, USP, Multi-Compendial, GMP, J.T.Baker™
(±)-1,2-Propanediol, ≥99.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

(±)-1,2-Propanediol, ≥99.5%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 L
19 L
Catalog number 9403-03
Price (USD)
-
Request A Quote
Quantity:
4 L
This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
Specifications
Boiling Point187.6°C (1013 hPa)
CAS57-55-6
CAS57-55-6
Chemical Name or Material(±)-1,2-Propanediol
ColorColorless
Melting Point-59°C
Molecular FormulaCH3CH(OH)CH2OH
PackagingAmber glass bottle, narrow mouth
SynonymPropylene glycol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, 1,2-Propanediol, (±)-1,2-Dihydroxy propane
GradeMulti-Compendial
Molecular Weight (g/mol)76.1
Percent Purity≥99.5%
Quantity4 L
Unit SizeEach