Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
Finntip™ Stepper Pipette Tips
Thermo Scientific™
Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
940423350 mLSterile
9404170500 μLNon-sterile
9404173500 μLSterile
94041801250 μLNon-sterile
94041831250 μLSterile
94041902.5 mLNon-sterile
94041932.5 mLSterile
94042005 mLNon-sterile
94042035 mLSterile
940421012.5 mLNon-sterile
940421312.5 mLSterile
940422025 mLNon-sterile
940422325 mLSterile
940423050 mLNon-sterile
Catalog number 9404233
Price (USD)
26.91
Online exclusive
29.90
Save 2.99
Case of 10
Volume (Metric):
50 mL
Sterility:
Sterile
Price (USD)
26.91
Online exclusive
29.90
Save 2.99
Case of 10
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Certifications/ComplianceISO 9001, ISO 14001
Volume (Metric) Dispensing1000, 2000, 3000, 4000, 5000 μL
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific FinnPipette Stepper
FormatIndividually Wrapped
Product LineFinnTip
SterilitySterile
Volume (Metric)50 mL
Unit SizeCase of 10