Finntip™ Filtered Pipette Tips
Thermo Scientific™

Finntip™ Filtered Pipette Tips

Protect pipettes and the samples from contamination with these filtered pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)
940524301000 μL10.5 cm
9405200010 μL3.2 cm
9405202020 μL3.3 cm
9405206050 μL4.8 cm
9405210010 μL5.2 cm
9405215020 μL5.2 cm
9405216030 μL5.2 cm
94052200100 μL5.2 cm
94052300200 μL5.2 cm
94052310100 μL7.8 cm
94052320200 μL7.8 cm
94052350300 μL5.2 cm
940524101000 μL7.1 cm
9405280010 mL15 cm
94052450150 to 1500 μL6.99 cm
940525505 mL14.7 cm
Catalog number 94052430
Price (USD)
236.47
Online exclusive
285.00
Save 48.53
Case of 480
Volume (Metric):
1000 μL
Length (Metric):
10.5 cm
Price (USD)
236.47
Online exclusive
285.00
Save 48.53
Case of 480

Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0.2μL-10mL.

Key Features:

  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Filtered Finntip pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
  • Inert and non-sealing polyethylene filter
  • Tips are supplied in vacuum sealed, sterilized rack

Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Versatile Tip Range

  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 20, 30, 100, 200, 300 and 1000 μL
  • Extended length tips available for volumes 100, 200 and 1000 μL allowing for better reach with specific labware consumables
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL

Immediate traceability

  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert

Note:

Availability may vary by country.

Specifications
DescriptionFinntip Filter 1000 μL Ext, sterile
DisposableSingle-use
Length (Metric)10.5 cm
AutoclavableNot Autoclavable
Color CodeBlue
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Thermo Scientific Finnpipette
FormatRack
Product LineFinnTip Filtered
SterilitySterile
Volume (Metric)1000 μL
Unit SizeCase of 480