Finntip™ Flex Filtered Pipette Tips - FAQs
View additional product information for Finntip™ Flex Filtered Pipette Tips - FAQs (94056810, 94056710, 94056580, 94056380, 94056980, 94056510, 94056520, 94056970)
1 product FAQs found
Can I autoclave Finntip Flex Filtered Pipette Tips and the rack they come in?
No, the Finntip Flex Filtered Pipette Tips cannot be autoclaved. However, the empty racks can be autoclaved. We recommend using the following autoclave conditions for PolyPropylene material: 121 degrees C, 1 bar for 20 minutes.