Finntip™ Flex Filtered Pipette Tips - FAQs
View additional product information for Finntip™ Flex Filtered Pipette Tips - FAQs (94056810, 94056710, 94056580, 94056380, 94056980, 94056510, 94056520, 94056970)
Error:This product does not have any FAQs available.