Product Image
Thermo Scientific™
Capillary Tubing for iCE™ 3000 Series AAS instruments
For best results, use only high quality, dedicated consumables, such as this Thermo Scientific™ Capillary Tubing for iCE™ 3000 Series AAS instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
9423390054210.5 mm ID
9423390054110.4 mm ID
Catalog number 942339005421
Price (USD)
13.00
Each
Diameter (Metric):
0.5 mm ID
Price (USD)
13.00
Each
For best results, use only high quality, dedicated consumables, such as this Thermo Scientific™ Capillary Tubing for iCE™ 3000 Series AAS instruments.
Choose consumables to fit your needs. Capillary tubing is available as:
  • 0.4mm ID
  • 0.5mm ID
Specifications
Diameter (Metric)0.5 mm ID
For Use With (Equipment)iCE 3000 Series AAS Instruments
TypeCapillary Tubing
Unit SizeEach