Product Image
Thermo Scientific™
STAT (Slotted Tube Atom Trap) Accessory for the iCE™ 3000 Series Flame AA Spectrometers
Improve precison and mimimize interferences compared to conventional Flame AAS analysis by adding a Thermo Scientific™ STAT to your Flame AAS instrument.
Have Questions?
Catalog NumberType
942339035011Slotted Tube Atom Trap
Catalog number 942339035011
Price (USD)
835.00
Each
Type:
Slotted Tube Atom Trap
Price (USD)
835.00
Each
Enhance the performance of the Thermo Scientific™ iCE™ 3000 Series Flame AA spectrometers with the Thermo Scientific™ STAT (Slotted Tube Atom Trap) Accessory.
By adding a STAT accessory to your Flame AAS instrument, precision is improved and interferences are minimized, as compared to conventional Flame AAS analysis.
Product Requirements
Requires the use of a Thermo Scientific™ 50mm Universal Burner, as included on the Thermo Scientific™ iCE™ 3300 Flame AAS instrument
Specifications
DescriptionStat (Requires Use Of 50 mm Universal Burner)
For Use With (Equipment)iCE 3000 Series Flame AA Spectrometers
TypeSlotted Tube Atom Trap
Unit SizeEach