Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
ClipTip™ 384-Format Pipette Tips
ClipTip™ 384-Format Pipette Tips
ClipTip™ 384-Format Pipette Tips
ClipTip™ 384-Format Pipette Tips
ClipTip™ 384-Format Pipette Tips
Thermo Scientific™

ClipTip™ 384-Format Pipette Tips

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityTip Style
9441005012.5 μLNon-sterileStandard 384
9441007312.5 μLSterileExtended Length
9441007012.5 μLNon-sterileExtended Length
9441005312.5 μLSterileStandard 384
9441010030 μLNon-sterileStandard 384
9441010330 μLSterileStandard 384
94410150125 μLNon-sterileStandard 384
94410153125 μLSterileStandard 384
Catalog number 94410050
Price (USD)
357.30
Online exclusive
397.00
Save 39.70
Case of 3840
Volume (Metric):
12.5 μL
Sterility:
Non-sterile
Tip Style:
Standard 384
Price (USD)
357.30
Online exclusive
397.00
Save 39.70
Case of 3840

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovativesnap and seal tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.

Key Features:

  • All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
  • Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
  • Ideal for 384-format microplate applications

Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Versatile Tip Range

  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables

Immediate traceability

  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert

Note:

Availability may vary by country.

Specifications
DescriptionClipTip 384 12.5, Non-filtered, Non-sterile, Rack
Length (English)1.18 in.
Length (Metric)3.0cm
Packaging10 x 384 tips/rack
AutoclavableNot Autoclavable
Color CodePink
FilteredNon-filtered
Fit for PurposePCR Ready
For Use With (Equipment)Thermo Scientific F1-ClipTip, Thermo Scientific E1-ClipTip
FormatRack
Product LineClipTip
SterilityNon-sterile
Tip StyleStandard 384
Volume (Metric)12.5 μL
Unit SizeCase of 3840