You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Pipette Products
▸
Pipette Tips
▸
Pipette Specific Tips
▸
ClipTip™ Pipette Tip Racks
Actual product may vary
Thermo Scientific™
ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Have Questions?
Change view
Catalog Number
Description
94410219
ClipTip Empty Rack, Small
94410619
ClipTip Empty Rack
94410519
ClipTip Empty Rack, Medium
94410819
ClipTip Empty Rack, Large
4 Options
Catalog number
94410219
Price (MXN)
-
Description:
ClipTip Empty Rack, Small
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Small Rack: For use with ClipTip 20 and 50 Pipette Tips
Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
Description
ClipTip Empty Rack, Small
Model
ClipTip Empty Rack
For Use With (Equipment)
Thermo Scientific E1-ClipTip
Product Line
ClipTip
Unit Size
Case of 10