You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Pipette Products
▸
Pipette Tips
▸
Pipette Specific Tips
▸
ClipTip™ Pipette Tip Racks
Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual.
Compare options for filtered tips »
Actual product may vary
Thermo Scientific™
ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Have Questions?
Change view
Catalog Number
Description
94410519
ClipTip Empty Rack, Medium
94410219
ClipTip Empty Rack, Small
94410619
ClipTip Empty Rack
94410819
ClipTip Empty Rack, Large
4 Options
Catalog number
94410519
Price (USD)
30.27
Online exclusive
35.20
Save 4.93
Case of 10
Add to cart
Description:
ClipTip Empty Rack, Medium
Price (USD)
30.27
Online exclusive
35.20
Save 4.93
Case of 10
Add to cart
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Small Rack: For use with ClipTip 20 and 50 Pipette Tips
Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
Description
ClipTip Empty Rack, Medium
Model
ClipTip Empty Rack
For Use With (Equipment)
ClipTip 1000 Pipette Tip
Product Line
ClipTip
Unit Size
Case of 10