ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
Thermo Scientific™

ClipTip™ Filtered 384-Format Pipette Tips

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip Style
9442005312.5 μLStandard 384
9442007312.5 μLExtended Length
9442010330 μLStandard 384
94420153125 μLStandard 384
Catalog number 94420053
Price (USD)
572.40
Online exclusive
636.00
Save 63.60
Case of 3840
Volume (Metric):
12.5 μL
Tip Style:
Standard 384
Price (USD)
572.40
Online exclusive
636.00
Save 63.60
Case of 3840

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovativesnap and seal tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.

Key Features:

  • All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
  • Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
  • Ideal for 384-format microplate applications
  • Filtered tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations

Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Versatile Tip Range

  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended length tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables

Immediate traceability

  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert

Note:

Availability may vary by country.

Specifications
DescriptionClipTip 384 12.5, Filtered, Sterile, Rack
Length (English)1.2 in.
Length (Metric)3.0cm
Packaging10 x 384 tips/rack
AutoclavableNot Autoclavable
Color CodePink
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip
FormatRack
Product LineClipTip
SterilitySterile
Tip StyleStandard 384
Volume (Metric)12.5 μL
Unit SizeCase of 3840