Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Thermo Scientific™

Immuno Breakable Modules Clear

Choose from four surfaces and a variety of wells and configurations. Immuno Breakable Modules offer an optimal choice of products for the specific assay.
Have Questions?
Catalog NumberBinding Property
95029180Hydrophilic
Catalog number 95029180
Price (USD)
440.31
Online exclusive
513.59
Save 73.28
Case of 50
Binding Property:
Hydrophilic
Price (USD)
440.31
Online exclusive
513.59
Save 73.28
Case of 50

Choose from four surfaces and a variety of wells and configurations. Thermo Scientific™ Immuno Breakable Modules offer an optimal choice of products for the specific assay. They offer lot-to-lot reliability and well-to-well reproducibility.

Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
  • For solid phase immuno assays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fit standard equipment
  • High optical quality for reliable results
  • Polystyrene

Choice of Surface

  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Binding PropertyHydrophilic
CertifiedYes
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
For Use With (Application)For antibody sandwich assays
FormatStrip Plate
LidWithout Lid
No. per Case50
No. per Pack50
Packaging25 Pk., 50 Cases
Plate BlockingNone
Product TypeStrip Plate
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Well400 μL
Volume (Metric) Working350 μL
Well DesignF8
ColorClear
MaterialPolystyrene
No. of Wells96
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.