Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Thermo Scientific™

Orion™ Carbon Dioxide Combination Ion Selective Electrode

Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly, accurately and economically.
Have Questions?
Catalog NumberConnector Types
9502BNWPBNC (Waterproof)
Catalog number 9502BNWP
Price (USD)
1,143.00
Online exclusive
1,270.00
Save 127.00
Each
Connector Types:
BNC (Waterproof)
Price (USD)
1,143.00
Online exclusive
1,270.00
Save 127.00
Each

Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately with the Thermo Scientific™ Orion™ 9502BNWP Carbon Dioxide Electrode. 

Features:

  • It features a gas-permeable membrane technology for use with loose membranes and O-rings.

Solutions required:

  • Standard
    • 950206 0.1 M NaHCO3 Carbon Dioxide standard, 475 mL
    • 950207 1000 ppm as CaCO3 Carbon Dioxide standard, 475 mL
  • Carbon dioxide buffer solution
    • 950210 Carbon Dioxide Ionic Strength Adjuster (ISA), 475 mL
  • Electrode fill solution
    • 950202 Carbon Dioxide electrode fill solution, 60 mL
  • Storage solution
    • 941706 0.1 M NaCl Chloride standard, 475 mL

Items included with:

  • 950202 Carbon Dioxide electrode fill solution, 60 mL 1 bottle
  • 950204 4 membranes with 1 O-ring

Warranty period:

  • 12 months from date of purchase
Specifications
DescriptionCarbon Dioxide Electrode
Connector TypesBNC (Waterproof)
For Use With (Equipment)ISE Meter with a BNC Connection
JunctionAnnular PTFE
MaterialEpoxy
Temperature (Metric) Operating0°C to 50°C
TypeCarbon Dioxide Electrode
Unit SizeEach