Microplate Sealing Tape - FAQs
Microplate Sealing Tape - FAQs
View additional product information for Microplate Sealing Tape - FAQs (9503130)
1 product FAQs found
Do you offer Microplate Sealing Tape, Cat. No. 9503130?
Yes, you may purchase it from Fisher Scientific (https://www.fishersci.ca/shop/products/thermo-scientific-microplate-sealing-tape/21377513).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.