3UV Lamp (115V, 60Hz)
Thermo Scientific™

3UV Lamp (115V, 60Hz)

Thermo Scientific Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252nm, 302nm and 365nm dial-selectable wavelength settings especially forRead more
Have Questions?
Catalog NumberQuantity
950341 unit
Catalog number 95034
Price (KRW)
1,708,000
Each
Quantity:
1 unit
Price (KRW)
1,708,000
Each

Thermo Scientific Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252nm, 302nm and 365nm dial-selectable wavelength settings especially for use with photoactivatable crosslinkers and UV-crosslinking methods. This 115 V, 60 Hz lamp is designed for grounded outlets in the USA.

Featuresof Pierce UV Lamps:

  • Multifunctional – three lamps (254nm, 302nm, 365nm) selected by a turn of the dial
  • Efficient – unique reflector design provides maximum UV for all applications
  • Lightweight and ergonomic – designed for hand-held or stationary use
  • Small footprint – 15.5' Length x 3' Width x 4.5' Depth (394mm x 76mm x 114mm)
  • Durable – lamp features aluminum construction and scratch-resistant finish
  • Versatile – 8-watt lamps are ideal for a wide area of illumination, such as culture plates
  • Validated for crosslinking – our researchers have validated this lamp for use in crosslinking applications with diazirine and phenyl azide photoactivatable crosslinkers

These 3UV lamp from UVP is a compact ultraviolet lamp for longwave, midrange and shortwave UV applications. The lamp's innovative design allows you to dial-in the required wavelength with just the turn of a knob. The lamp is compatible with our extensive line of photo-reactive crosslinking and label-transfer reagents, including the Photo-reactive Amino Acids for discovering in-cell protein interactions.

Applications:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
DescriptionPierce 3UV Lamp, 115 Volts, 60 Hz (USA)
Quantity1 unit
Light SourceUV
Product TypeTriple Wavelength UV Lamp
Voltage115 V
Wattage8 W
Wavelength365/302/254 nm
Unit SizeEach

Frequently asked questions (FAQs)

Do you offer replacement bulbs for 3UV Lamp (115V, 60Hz) (Cat. No. 95034)?

The catalog numbers for replacement bulbs for 3UV Lamp (115V, 60Hz) are as follows:

  • Short wave: 34-0013-01
  • Long wave 34-0034-01
  • Mid-range 34-0044-01