KingFisher™ Plastics for 24 deep-well format
Thermo Scientific™

KingFisher™ Plastics for 24 deep-well format

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
9504047024 Deep-well PlateKF Duo Prime, Flex, Presto
9700258024 Combi tip combKingFisher Apex
9504048024 Deep-well Plate, SterileKF Duo Prime, Flex, Presto
9700261024 DW tip comb and plateKingFisher Flex and Presto
9700262024 DW tip comb, plate, sterileKingFisher Flex and Presto
970035106-tip comb and 24 DW plateKingFisher Duo Prime
95040470B24 Deep Well plate, barcodedKingFisher Apex
97002610B24 DW tip comb, plate, barcodeKingFisher Apex
95040480B24 DW plate, barcoded, sterileKingFisher Apex
97002620B24 DW tip comb, plate, sterileKingFisher Apex
Catalog number 95040470
Price (USD)
336.65
Online exclusive
411.00
Save 74.35
Case of 50
Product Type:
24 Deep-well Plate
For Use With (Equipment):
KF Duo Prime, Flex, Presto
Price (USD)
336.65
Online exclusive
411.00
Save 74.35
Case of 50

Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.

Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

For research use only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KF Duo Prime, Flex, Presto
Capacity24 Samples
Format24 Deep-well Plate
Product Type24 Deep-well Plate
Unit SizeCase of 50

Frequently asked questions (FAQs)

What is the advantage of using the KingFisher Flex instrument with the 24 Deep-Well magnetic head?

The KingFisher Flex instrument with the 24 Deep-Well magnetic head can process larger volumes for each sample (up to 5mL total volume) compared to the KingFisher Flex instrument with the 96 Deep-Well magnetic head.

Can I use the same tips and plates for the KingFisher Flex instrument with the 96-well magnetic head and the MagMAX Express-96 instrument magnetic head?

Yes, the KingFisher Flex instrument 96-well magnetic head and the MagMAX Express-96 instrument magnetic head can use the same tip combs and KingFisher plates even though the magnetic heads are different.

What is the quantity provided for KingFisher 24 deep-well plate (for Duo Prime, Flex and Presto) (Cat. No. 95040470)?

The quantity is 50.

I see that the KingFisher 24 Deep Well Head has a processing volume of 5 mL. Does that mean that I can process up to 5 mL of my sample?

No, the processing volume refers to the maximum volume that can be held in the 24 Deep Well plates. This incorporates the volume of binding buffers and beads, so when scaling up reaction volumes, it is important to make sure that the total volume does not exceed 5 mL.

My KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?

No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original KingFisher Flex script would have to be created that starts off at the point where the run was stopped.