Orion™ Gas-Sensing ISE membranes
Thermo Scientific™
Orion™ Gas-Sensing ISE membranes
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Includes
951205Standard Ammonia Electrode, 9512BNWP, 95123 Bonded Membranes
950204Carbon Dioxide Electrode, 9502BNMP4 Membranes
951204Standard Ammonia Electrode, 9512BNWP, 951220 Loose Membranes
951214HP Ammonia Electrode, 9512HPBNWP, 9512HP20 Loose Membranes
Catalog number 951205
Price (USD)
226.80
Online exclusive
252.00
Save 25.20
Pack of 3
For Use With (Equipment):
Standard Ammonia Electrode, 9512BNWP, 9512
Includes:
3 Bonded Membranes
Price (USD)
226.80
Online exclusive
252.00
Save 25.20
Pack of 3
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionGas-Sensing ISE membrane
Includes3 Bonded Membranes
For Use With (Equipment)Standard Ammonia Electrode, 9512BNWP, 9512
TypeGas-Sensing ISE Membrane
Unit SizePack of 3