CarrierMax™ SMN1/SMN2 Reagent Kit
Applied Biosystems™

CarrierMax™ SMN1/SMN2 Reagent Kit

The CarrierMax SMN1/SMN2 Reagent Kit uses multiplex PCR amplification of genomic DNA and capillary electrophoresis to detect deletions in exonRead more
Have Questions?
Catalog NumberQuantity
95236350 Samples
Catalog number 952363
Price (USD)
-
Quantity:
50 Samples
The CarrierMax SMN1/SMN2 Reagent Kit uses multiplex PCR amplification of genomic DNA and capillary electrophoresis to detect deletions in exon 7 of the Survival of Motor Neuron 1 (SMN1) gene that result in truncated SMN protein. In addition, the kit detects two variants associated with the silent carrier (2+0) haplotype. The results are processed with GeneMapper Software in conjunction with included CarrierMAX Software to consolidate data and simplify carrier classifications in spinal muscular atrophy (SMA) carrier screening research.

Key features of the kit include the ability to:
• Accurately and consistently detect Exon 7 copy number states of 0, 1, or ≥ 2 for both SMN1 and SMN2
• Determine if SMN1 to SMN2 Exon 7 gene conversion has occurred
• Detect haplotype variants associated with SMA silent (2+0) carrier status
• Perform fragment analysis on the Applied Biosystems 3500/3500xL Genetic Analyzer or SeqStudio Genetic Analyzer
• Analyze the results in GeneMapper Software, followed by easy reporting of sample carrier classification in the included CarrierMax Software

SMA associated with loss of SMN1 is one of the leading causes of neurodegenerative disorders and is a critical target for carrier screening in individuals planning for conception. SMA arises from the loss of SMN1 gene through genetic disruptions including point or structural mutations. The majority (95%) of SMN1 losses are due to complete deletion of the gene or a point mutation on Exon 7 that causes an SMN1 splice variant to occur and results in a less functional SMN2 transcript. In some cases, genetic recombination results in two SMN1 genes on a single chromosome and are missed as carriers (2+0) using conventional methods. The CarrierMax SMN1/SMN2 Reagent Kit offers the ability to quickly and accurately screen many samples to determine the SMN1 Exon 7 copies present in the sample along with indication of potential 2+0 carriers through haplotype assessment.

How to use
The kit comes with enough reagents to run 50 samples for SMN1 carrier screening research on either the SeqStudio or Applied Biosystems 3500 genetic analyzers. The premise behind the SMN1 Exon 7 copy number determination is the use of a dual PCR based system assessing both copy number and key variants.

The optimized workflow involves preparation of high quality DNA from a sample source (e.g., whole blood), dual PCR cycling reactions (∼2 hours), and subsequent CE run (∼1 hour) and data analysis (∼10 min).

Download CarrierMax Software (.zip) ›
SeqStudio GeneMapper files
3500/XL GeneMapper files

Quality of performance
The analytical sensitivity and specificity of the assay is proven to be >99% using Coriell control standards. Accuracy was determined to be >99% in carrier classification of reference samples. Reliability of the system as determined by reproducible classifications across experiments, labs, and systems indicates the suitability of the CarrierMax SMN1/SMN2 kit for carrier screening applications such as Expanded Carrier Screening research.

Specifications
For Use With (Application)Genotyping Using Fragment Analysis for Carrier Screening Research
For Use With (Equipment)Applied Biosystems 3500 Genetic Analyzer, Applied Biosystems 3500 XL Genetic Analyzer, Applied Biosystems SeqStudio Genetic Analyzer
Label or Dye6-FAM, HEX, ROX
Product TypeSMN1/SMN2 Reagent Kit
Quantity50 Samples
TargetSMN1, SMN2
FormatKit
Unit SizeEach

Frequently asked questions (FAQs)

Do you have GeneMapper analysis parameters available for the CarrierMax SMN1/SMN2 Reagent Kit?

We have GeneMapper analysis parameters for the CarrierMax kits, specifically for the SeqStudio and the 3500/3500xL instruments. They can be found on the following links or the CarrierMax kit product page. http://downloads.thermofisher.com/GeneMapper/FMR1_SeqStudio_GeneMapper_files.zip http://downloads.thermofisher.com/GeneMapper/FMR1_3500_GeneMapper_files.zip

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What software can I use to analyze data from the CarrierMax SMN1/SMN2 Reagent Kit?

To analyze data from the CarrierMax SMN1/SMN2 Reagent Kit, you would have to use both the GeneMapper Software and CarrierMax Software for genotyping and classification of the sample, respectively.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Is the CM500 size standard available separately from the CarrierMax SMN1/SMN2 Reagent Kit?

The CM500 size standard found in the CarrierMax SMN1/SMN2 Reagent Kit is not available separately.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What size standard is used in the CarrierMax SMN1/SMN2 Reagent Kit?

The CarrierMax SMN1/SMN2 Reagent Kit uses the CM 500 size standard.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Which fluorescent dye labels are used in the CarrierMax SMN1/SMN2 Reagent Kit?

The CarrierMax SMN1/SMN2 Reagent Kit uses 6-FAM and HEX fluorescent dye labels for the primers and the ROX fluorescent dye label for the size standard.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.