Do you offer an alternative for the discontinued DMET PLUS Assay (Cat. No. 901268)?
We recommend reviewing the Microarrays for Preemptive Pharmacogenomics page to determine a potential alternative.
If you have any questions you can email Tech Support at techsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
If you have any questions you can email Tech Support at techsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.