Especificações
Boiling Point118°C (1013 hPa)
CAS64-19-7
Chemical Name or MaterialAcetic Acid, glacial
ColorColorless
Melting Point17°C
Molecular FormulaH3CCOOH
PackagingClear glass bottle
SynonymMethanecarboxylic acid, Glacial acetic acid, Vinegar acid, Ethanoic acid
GradeMulti-Compendial/NF
Molecular Weight (g/mol)60.05
Percent Purity99.5-100.05%
Quantity2.5 L
Unit SizeEach