Acetic acid, glacial, 99.5-100.05%, USP, Multi-Compendial, GMP Manufactured Product, J.T.Baker™
Avantor™

Acetic acid, glacial, 99.5-100.05%, USP, Multi-Compendial, GMP Manufactured Product, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
15 L
450 lb.
Packaging:
View all
Polyethylene drum
Catalog number 9526-20
Price (MXN)
-
Quantity:
450 lb.
Packaging:
Polyethylene drum
This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
Specifications
Boiling Point118°C (1013 hPa)
CAS64-19-7
Chemical Name or MaterialAcetic Acid, glacial
ColorColorless
Melting Point17°C
Molecular FormulaH3CCOOH
PackagingPolyethylene drum
SynonymMethanecarboxylic acid, Glacial acetic acid, Vinegar acid, Ethanoic acid
GradeMulti-Compendial/NF
Molecular Weight (g/mol)60.05
Percent Purity99.5-100.05%
Quantity450 lb.
Unit SizeEach