Automation Tips for Qiagen™ Workstations
Thermo Scientific™
Automation Tips for Qiagen™ Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredPackagingFit for Purpose
953-2611100 μLNon-filtered96 Tips/Tray, 10 Trays/CaseSterile
951-021250 μLFiltered96 Tips/Tray, 10 Trays/CaseSterile and PCR Ready
951-253300 μLNon-filtered96 Tips/Tray, 50 Trays/CaseSterile
951-261300 μLNon-filtered96 Tips/Tray, 10 Trays/CaseSterile
951-262300 μLNon-filtered96 Tips/Tray, 10 Trays/CaseCertified Quality
953-0211000 μLFiltered96 Tips/Tray, 10 Trays/CaseSterile and PCR Ready
953-2531100 μLNon-filtered96 Tips/Tray, 50 Trays/CaseSterile
953-2621100 μLNon-filtered96 Tips/Tray, 10 Trays/CaseCertified Quality
Catalog number 953-261
Price (USD)
126.03
Online Exclusive
177.00
Save 50.97 (29%)
Case of 960
Volume (Metric):
1100 μL
Filtered:
Non-filtered
Packaging:
96 Tips/Tray, 10 Trays/Case
Fit for Purpose:
Sterile
Price (USD)
126.03
Online Exclusive
177.00
Save 50.97 (29%)
Case of 960
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:: A five (5)-year shelf life from the date of manufacture is recommended.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Packaging96 Tips/Tray, 10 Trays/Case
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Universal Fit, Qiagen
FormatRack
Product LineQiagen
SterilitySterile
Tip StyleAutomation
Volume (Metric)1100 μL
Unit SizeCase of 960