Automation Tips for Qiagen™ Workstations
Thermo Scientific™
Automation Tips for Qiagen™ Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)FilteredPackagingFit for Purpose
953-2621100 μLNon-filtered96 Tips/Tray, 10 Trays/CaseCertified Quality
951-011250 μLFiltered96 Tips/Tray, 10 Trays/CaseSterile and PCR Ready
951-021250 μLFiltered96 Tips/Tray, 10 Trays/CaseSterile and PCR Ready
951-253300 μLNon-filtered96 Tips/Tray, 50 Trays/CaseSterile
951-261300 μLNon-filtered96 Tips/Tray, 10 Trays/CaseSterile
951-262300 μLNon-filtered96 Tips/Tray, 10 Trays/CaseCertified Quality
953-0211000 μLFiltered96 Tips/Tray, 10 Trays/CaseSterile and PCR Ready
953-2531100 μLNon-filtered96 Tips/Tray, 50 Trays/CaseSterile
953-2611100 μLNon-filtered96 Tips/Tray, 10 Trays/CaseSterile
Número do catálogo 953-262
Preço (BRL)
-
Volume (Metric):
1100 μL
Filtered:
Non-filtered
Packaging:
96 Tips/Tray, 10 Trays/Case
Fit for Purpose:
Certified Quality
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:: A five (5)-year shelf life from the date of manufacture is recommended.
Especificações
Packaging96 Tips/Tray, 10 Trays/Case
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Universal Fit, Qiagen
FormatRack
Product LineQiagen
SterilityNon-sterile
Tip StyleAutomation
Volume (Metric)1100 μL
Unit SizePack of 960