What is the concentration of the TaqMan SARS-CoV-2, Flu A, Flu B RNA Control material in the TaqMan SARS-CoV-2, Flu A, Flu B RT-PCR Assay Kit?
The TaqMan SARS-CoV-2, Flu A, Flu B RNA Control concentration is proprietary. This control is intended to be used as a process quality control with the TaqMan SARS-CoV-2, Flu A, Flu B Multiplex Assay. It is designed to confirm that viral targets can be routinely detected at low target levels and should not be used for workflow validation purposes.