Nitric acid, 69.0-70.0%, NF, GMP, J.T.Baker™
Avantor™

Nitric acid, 69.0-70.0%, NF, GMP, J.T.Baker™

This product is GMP Manufactured and meets N.F. Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Clear glass bottle, PVC-coated
Clear glass bottle
Catalog number 9607-33
Price (USD)
-
Quantity:
2.5 L
Packaging:
Clear glass bottle, PVC-coated
This product is GMP Manufactured and meets N.F. Requirements.
This product is GMP Manufactured and meets N.F. Requirements.
Specifications
Boiling Point120.5°C (1013 hPa)
CAS7697-37-2
Chemical Name or MaterialNitric acid
ColorColorless to slightly yellow
Melting Point-42°C
Molecular FormulaHNO3
PackagingClear glass bottle, PVC-coated
SynonymAqua Fortis, Azotic Acid
GradeNF
Molecular Weight (g/mol)63.01
Percent Purity69.0-70.0%
Quantity2.5 L
Unit SizeEach