Sulfuric acid, 95.0-98.0% (by acidimetry), NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sulfuric acid, 95.0-98.0% (by acidimetry), NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets N.F., EP, and BP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Catalog number 9671-03
Price (USD)
-
Quantity:
2.5 L

This product is GMP Manufactured and meets N.F., EP, and BP Requirements.

Specifications
Boiling Point∼335°C (1013 hPa)
CAS7664-93-9
Chemical Name or MaterialSulfuric acid
ColorColorless
Melting Point3°C
Molecular FormulaH2SO4
PackagingClear glass bottle
SynonymSulfuric acid
GradeMulti-Compendial/NF
Molecular Weight (g/mol)98.08
Percent Purity95.0-98.0%
Quantity2.5 L
Unit SizeEach