Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Thermo Scientific™
Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Measure lead ions in aqueous solutions quickly, simply, accurately and economically.
Have Questions?
Catalog NumberConnector Types
9682BNWPBNC (Waterproof)
Catalog number 9682BNWP
Price (USD)
1,341.00
Online exclusive
1,490.00
Save 149.00
Each
Connector Types:
BNC (Waterproof)
Price (USD)
1,341.00
Online exclusive
1,490.00
Save 149.00
Each
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
Features:
  • 9682BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.The lead electrode makes a sensitive endpoint detector for the titration of sulfate ions with lead perchlorate.
Solutions required:
  • Standard
    • 948206 0.1 M lead standard, 475 mL bottle
    • 948207 0.1 M sodium sulfate standard for sulfate analysis with the lead ISE, 475 mL bottle
  • Electrode fill solution
    • 900062 Optimum Results B electrode filling solution, 5 x 60 mL bottles
Items included with:
  • 900062 Optimum Results B electrode filling solution, 60 mL 1 bottle
  • 948201 Polishing strip. Qty 2.
Warranty period:
  • 12 months from date of purchase
Specifications
DescriptionWith waterproof BNC connector
Driftdrift-free potentials
Connector TypesBNC (Waterproof)
For Use With (Equipment)ISE Meter
JunctionSure-Flow Junction
TypeLead Ion Selective Electrode, BNC waterproof connector
Unit SizeEach