KingFisher™ Systems Consumables, KingFisher plate 200μL - FAQs
KingFisher™ Systems Consumables, KingFisher plate 200μL - FAQs
View additional product information for KingFisher™ Systems Consumables - FAQs (97002084, 97002080, 97002070)
1 product FAQs found
Are the MagMAX Express and KingFisher plates the same?
Yes, the MagMAX Express and KingFisher plates are equivalent. We recommend the KingFisher plates as an alternative to the discontinued MagMAX Express plates.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.