KingFisher™ Plastics for 96 deep-well format
Thermo Scientific™
KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
97002534BBarcoded Tip CombKingFisher Apex
97002570Tip CombKingFisher Apex
97002534Tip CombKingFisher Flex and Presto
97003500Tip CombKingFisher Duo Prime only
97003520Elution StripKingFisher Duo Prime only
97003540Elution Strip CapKingFisher Duo Prime only
97003530Combi PackKingFisher Duo Prime only
Catalog number 97002534B
Price (USD)
734.65
Online exclusive
918.00
Save 183.35
Case of 100
Product Type:
Barcoded Tip Comb
For Use With (Equipment):
KingFisher Apex
Price (USD)
734.65
Online exclusive
918.00
Save 183.35
Case of 100
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionKingFisher 96 Deep-well Tip Comb, Barcoded
For Use With (Equipment)KingFisher Apex
Product LineKingFisher
Quantity100/Cs.
Product TypeBarcoded Tip Comb
Unit SizeCase of 100
Frequently asked questions (FAQs)
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.