KingFisher™ Plastics for 96 deep-well format
Thermo Scientific™

KingFisher™ Plastics for 96 deep-well format

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
97002570Tip CombKingFisher Apex
97002534Tip CombKingFisher Flex and Presto
97003500Tip CombKingFisher Duo Prime only
97003520Elution StripKingFisher Duo Prime only
97003540Elution Strip CapKingFisher Duo Prime only
97003530Combi PackKingFisher Duo Prime only
97002534BBarcoded Tip CombKingFisher Apex
Catalog number 97002570
Price (USD)
699.65
Online exclusive
874.00
Save 174.35
Case of 100
Product Type:
Tip Comb
For Use With (Equipment):
KingFisher Apex
Price (USD)
699.65
Online exclusive
874.00
Save 174.35
Case of 100

Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.

Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
DescriptionKingFisher Apex 96 Combi tip comb (for Apex only)
DisposableSingle-use
For Use With (Equipment)KingFisher Apex
Product LineKingFisher
Quantity100/Pk.
Capacity96 Samples
Format96 DW Comb
Product TypeTip Comb
Unit SizeCase of 100

Frequently asked questions (FAQs)

Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Deep-Well Tip Combs (Cat. No. A31537)?

Yes, we recommend using KingFisher 96 tip combs for deep-well format (Cat. No. 97002534 or 97002570).

Are KingFisher Plastics for 96-deep well format autoclavable?

No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.

Can I reuse the Kingfisher Plastics for 96 deep-well format?

The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.

Are the KingFisher Plastics for 96 deep-well format CE marked?

The KingFisher Plastics for 96 deep-well format are not CE marked.