KingFisher™ Plastics for 24 deep-well format
KingFisher™ Plastics for 24 deep-well format
KingFisher™ Plastics for 24 deep-well format
KingFisher™ Plastics for 24 deep-well format
KingFisher™ Plastics for 24 deep-well format
Thermo Scientific™
KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
9700262024 DW tip comb, plate, sterileKingFisher Flex and Presto
9700258024 Combi tip combKingFisher Apex
9504047024 Deep-well PlateKF Duo Prime, Flex, Presto
9504048024 Deep-well Plate, SterileKF Duo Prime, Flex, Presto
9700261024 DW tip comb and plateKingFisher Flex and Presto
970035106-tip comb and 24 DW plateKingFisher Duo Prime
95040470B24 Deep Well plate, barcodedKingFisher Apex
97002610B24 DW tip comb, plate, barcodeKingFisher Apex
95040480B24 DW plate, barcoded, sterileKingFisher Apex
97002620B24 DW tip comb, plate, sterileKingFisher Apex
Catalog number 97002620
Price (USD)
1,082.65
Online exclusive
1,320.00
Save 237.35
Case of 50
Product Type:
24 DW tip comb, plate, sterile
For Use With (Equipment):
KingFisher Flex and Presto
Price (USD)
1,082.65
Online exclusive
1,320.00
Save 237.35
Case of 50
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
For research use only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher Flex and Presto
Capacity24 Samples
FormatPlate and Comb
Product Type24 DW tip comb, plate, sterile
Unit SizeCase of 50
Frequently asked questions (FAQs)
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What is the quantity provided for KingFisher 24 deep-well tip comb and plate (for Flex and Presto) (Cat. No. 97002610)?
The quantity is 50.
I see that the KingFisher 24 Deep Well Head has a processing volume of 5 mL. Does that mean that I can process up to 5 mL of my sample?
No, the processing volume refers to the maximum volume that can be held in the 24 Deep Well plates. This incorporates the volume of binding buffers and beads, so when scaling up reaction volumes, it is important to make sure that the total volume does not exceed 5 mL.
My KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?
No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original KingFisher Flex script would have to be created that starts off at the point where the run was stopped.
I have a 12-plate protocol but there are only 8 slots on the KingFisher Flex instrument turntable. What should I do with the remaining 4 plates?
Set up the protocol on the BindIt software. After the first 8 plates are done, the instrument will pause and instruct you to switch out the plates.