KingFisher 96 Deep-well Plates
Thermo Scientific™
KingFisher 96 Deep-well Plates
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)Content And Storage
97002820BTip Comb and Deep-well PlateKingFisher Apex
95040450Deep-well PlateKF Duo Prime, Flex and Presto
95040460Deep-well PlateKF Duo Prime, Flex and Presto
95040450BDeep-well PlateKingFisher Apex
95040450BUDeep-well PlateKingFisher Apex
95040460BDeep-well PlateKingFisher Apex
97002820Tip Comb and Deep-well PlateKingFisher Flex and Presto
Catalog number 97002820B
Price (USD)
815.65
Online exclusive
1,020.00
Save 204.35
Case of 50
Product Type:
Tip Comb and Deep-well Plate
For Use With (Equipment):
KingFisher Apex
Price (USD)
815.65
Online exclusive
1,020.00
Save 204.35
Case of 50
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Apex, KingFisher Flex, Duo Prime, and Presto instruments.
KingFisher™ Plastics for Automated DNA, RNA, and Protein Purification
KingFisher™ plastics consumables—including plates, strips, and tip combs—are specifically designed for automated magnetic bead-based purification of DNA, RNA, proteins, and cells. Engineered for easy compatibility with KingFisher Apex™, Flex™, Duo Prime, or Presto™ instruments, they enable reliable, high-throughput sample processing in research and clinical laboratories.
Purpose-Built for Magnetic Bead Workflows
Unlike standard lab plastics, KingFisher consumables are optimized for magnetic particle processing. Carefully designed plate formats, well geometries, and tip comb structures helps ensure efficient magnetic bead capture, transfer, and mixing—key factors that directly impact yield, purity, and reproducibility.
Tip combs protect magnetic rods while enabling precise bead handling, and plate designs support optimal reagent interaction throughout the purification workflow
Consistent Quality Through Controlled Manufacturing
Manufactured using high-purity, traceable raw materials in a fully automated, quality-controlled process, KingFisher plastics help deliver consistent performance across runs. This enables reliability in demanding applications where reproducibility is critical.
Each consumable is engineered to limit variability and support standardized workflows across laboratories.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher Apex
Product TypeTip Comb and Deep-well Plate
Quantity50/Cs.
BarcodeIncludes Barcode
No. of Wells96
Product LineKingFisher
SterilitySterile
Unit SizeCase of 50
Contents & Storage
KingFisher 96 Deep-well Tip Comb and Plate, sterile, Barcoded (for Apex)
Frequently asked questions (FAQs)
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.