KingFisher™ Plastics for 24 deep-well format
Thermo Scientific™
KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
970035106-tip comb and 24 DW plateKingFisher Duo Prime
9700258024 Combi tip combKingFisher Apex
9504047024 Deep-well PlateKF Duo Prime, Flex, Presto
9504048024 Deep-well Plate, SterileKF Duo Prime, Flex, Presto
9700261024 DW tip comb and plateKingFisher Flex and Presto
9700262024 DW tip comb, plate, sterileKingFisher Flex and Presto
95040470B24 Deep Well plate, barcodedKingFisher Apex
97002610B24 DW tip comb, plate, barcodeKingFisher Apex
95040480B24 DW plate, barcoded, sterileKingFisher Apex
97002620B24 DW tip comb, plate, sterileKingFisher Apex
Catalog number 97003510
Price (USD)
206.65
Online exclusive
252.00
Save 45.35
Each
Product Type:
6-tip comb and 24 DW plate
For Use With (Equipment):
KingFisher Duo Prime
Price (USD)
206.65
Online exclusive
252.00
Save 45.35
Each
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
For research use only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher Duo Prime
Quantity48 Pc.
FormatPlate and Comb
Product Type6-tip comb and 24 DW plate
Unit SizeEach
Frequently asked questions (FAQs)
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What kind of consumables do I need for the KingFisher Duo Purification System?
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
What is the difference between the KingFisher Duo and the KingFisher Duo Prime Purification Systems?
The KingFisher Duo Prime Purification System has the following additional features compared to the KingFisher Duo Purification System:
• Improved safety shield design
• UV lamp
• Handheld barcode reader option
• Additional language options
• Optimized elution strip block for better alignment of the plastic strip
The KingFisher Duo can run any script written for the KingFisher Duo Prime and does not require additional modifications.