KingFisher elution strip for 12 pin magnet (for Duo Prime only) - FAQs
KingFisher elution strip for 12 pin magnet (for Duo Prime only) - FAQs
View additional product information for KingFisher™ Plastics for 96 deep-well format - FAQs (97003520, 97003540, 97003530, 97002570, 97002534B, 97002534, 97003500)
5 product FAQs found
What kind of consumables do I need for the KingFisher Duo Purification System?
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
What is the difference between the KingFisher Duo and the KingFisher Duo Prime Purification Systems?
The KingFisher Duo Prime Purification System has the following additional features compared to the KingFisher Duo Purification System:
• Improved safety shield design
• UV lamp
• Handheld barcode reader option
• Additional language options
• Optimized elution strip block for better alignment of the plastic strip
The KingFisher Duo can run any script written for the KingFisher Duo Prime and does not require additional modifications.
Are KingFisher Plastics for 96-deep well format autoclavable?
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.
Can I reuse the Kingfisher Plastics for 96 deep-well format?
The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.
Are the KingFisher Plastics for 96 deep-well format CE marked?
The KingFisher Plastics for 96 deep-well format are not CE marked.