KingFisher Combi Pack for microtiter 96 deep-well plate (for Duo Prime only) - FAQs
KingFisher Combi Pack for microtiter 96 deep-well plate (for Duo Prime only) - FAQs
View additional product information for KingFisher™ Plastics for 96 deep-well format - FAQs (97003520, 97003540, 97003530, 97002570, 97002534B, 97002534, 97003500)
7 product FAQs found
How many plates come with the KingFisher combi pack for microtiter 96 deep-well plate (Cat. No. 97003530)?
This kit is for the KingFisher Duo Prime only. It contains the following for processing 96 samples:
• 8 plates
• 8 tip combs
• 8 elution strips
• 8 elution strip caps
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What kind of consumables do I need for the KingFisher Duo Purification System?
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
What is the difference between the KingFisher Duo and the KingFisher Duo Prime Purification Systems?
The KingFisher Duo Prime Purification System has the following additional features compared to the KingFisher Duo Purification System:
• Improved safety shield design
• UV lamp
• Handheld barcode reader option
• Additional language options
• Optimized elution strip block for better alignment of the plastic strip
The KingFisher Duo can run any script written for the KingFisher Duo Prime and does not require additional modifications.
Are KingFisher Plastics for 96-deep well format autoclavable?
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.
Can I reuse the Kingfisher Plastics for 96 deep-well format?
The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.
Are the KingFisher Plastics for 96 deep-well format CE marked?
The KingFisher Plastics for 96 deep-well format are not CE marked.