Orion™ Residual Chlorine Combination Ion Selective Electrode
Orion™ Residual Chlorine Combination Ion Selective Electrode
Orion™ Residual Chlorine Combination Ion Selective Electrode
Orion™ Residual Chlorine Combination Ion Selective Electrode
Orion™ Residual Chlorine Combination Ion Selective Electrode
Thermo Scientific™

Orion™ Residual Chlorine Combination Ion Selective Electrode

Measure total residual chlorine quickly and accurately.
Have Questions?
Catalog NumberConnector Types
9770BNWPBNC (Waterproof)
Catalog number 9770BNWP
Price (USD)
945.00
Online exclusive
1,050.00
Save 105.00
Each
Connector Types:
BNC (Waterproof)
Price (USD)
945.00
Online exclusive
1,050.00
Save 105.00
Each

Measure total residual chlorine quickly and accurately with Thermo Scientific™ Orion™ Chlorine Electrodes. All forms of total residual chlorine (free chlorine, hypochlorites and chlorine bound to nitrogenous compounds) are measured.

The ThermoFisher Orion Chlorine electrode complies with SM 4500-Cl I for Drinking water measurements.

Features:

  • The method permits rapid analysis to be done at the sampling site, eliminating erroneous low readings due to Cl2 loss. It requires the addition of two reagents to the sample before measurement.
  • Total chlorine residual in wastewater is measured directly by the Orion 9770BNWP Chlorine ion-selective electrode regardless of color or turbidity.

Solutions required:

  • Standards
    • 977007 100 ppm as Cl2 residual chlorine standard,75 mL bottle
  • Reagents
    • 977010 Iodide reagent, 5 x 50 mL bottles
    • 977011 Acid reagent, 475 mL bottle

Items included with:

  • 977010 Iodide reagent. 50 mL 1 bottle
  • 948201 Polishing strip. Qty 2

Warranty period:

  • 12 months from date of purchase
Specifications
DescriptionWaterproof BNC connector
Connector TypesBNC (Waterproof)
For Use With (Equipment)ISE Meter
JunctionSure-Flow Junction
MaterialEpoxy
Temperature (Metric) Operating0°C to 40°C
TypeCombination ISE
Unit SizeEach