NERL™ High Purity Water
Thermo Scientific™

NERL™ High Purity Water

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 gal.
5 gal.
Catalog number 9805
Price (USD)
-
Quantity:
5 gal.
Request bulk or custom format
Specifications
Recommended Storage15° to 30°C
Shelf Life12 Months
GradeNERL
Quantity5 gal.
pH5 to 7
View more
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.

Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.

Contains no preservatives

Ordering Information:

Certificate of analysis available for each case

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.

For In Vitro Diagnostic Use.