KingFisher™ Pure DNA Plant Kit - FAQs
KingFisher™ Pure DNA Plant Kit - FAQs
View additional product information for KingFisher™ Pure DNA Plant Kit - FAQs (98050496)
2 product FAQs found
Are the KingFisher Pure kits discontinued?
All KingFisher Pure kits are discontinued. We recommend using the MagMAX kits which are optimized for use with the KingFisher instruments. Please contact our Technical Support team at techsupport@thermofisher.com to find out which MagMAX kit we recommend as an alternative to the KingFisher Pure Kits.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What alternatives do you recommend for the discontinued KingFisher Pure kits?
Please contact our Technical Support team at techsupport@thermofisher.com to find out which MagMAX kits we recommend as an alternative to the KingFisher Pure Kits.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.