Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, Capacity: 17.3 cu. ft. (489.9L) capacity, double door, holds 288 boxes, 208-230V/60Hz, 12A, painted galvaneel interior - FAQs
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, Capacity: 17.3 cu. ft. (489.9L) capacity, double door, holds 288 boxes, 208-230V/60Hz, 12A, painted galvaneel interior - FAQs
View additional product information for Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (988)