Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, 120V/60Hz, 16A, painted galvaneel interior - FAQs
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, 120V/60Hz, 16A, painted galvaneel interior - FAQs
View additional product information for Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (990)