Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, Capacity: 13 cu. ft. (368.1L) capacity, double door, holds 216 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, Capacity: 13 cu. ft. (368.1L) capacity, double door, holds 216 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
View additional product information for Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (991)