Sterilin™ Caps for Glass Tubes
Actual product may vary
Thermo Scientific™
Sterilin™ Caps for Glass Tubes
Secure your samples with these autoclavable screw-thread caps for glass tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure SizeLiner MaterialNo. per Case
9998-1516 mmPTFE288
99999-1516 mmRubber1000
99999-1820 mmRubber1000
9998-1313 mmPTFE288
9998-1820 mmPTFE192
Catalog number 9998-15
Price (USD)
-
Closure Size:
16 mm
Liner Material:
PTFE
No. per Case:
288
Secure your samples with these autoclavable screw-thread caps for glass tubes. Thermo Scientific™ Sterilin™ Caps for Glass Tubes are available with choice of rubber or PTFE liner.
NOT AVAILABLE IN AMERICA
  • Phenolic caps to fit screw neck tubes 99449 and 99448
  • Autoclavable at 121°C
  • Choice of rubber of PTFE liner
Ordering Information
  • Available in UK only
Specifications
Closure Size16 mm
For Use With (Equipment)Glass Tubes
Liner MaterialPTFE
No. per Case288
AutoclavableAutoclavable
MaterialPhenolic
SterilityNon-sterile
Unit SizeEach